missing translation for 'onlineSavingsMsg'
Learn More

2-Ethyl-3-(methylthio)pyrazine 98.0+%, TCI America™

Catalog No. E07195G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E07195G 5 g
E071925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E07195G Supplier TCI America Supplier No. E07195G
Only null left

Chemical Identifiers

CAS 72987-62-3
Molecular Formula C7H10N2S
Molecular Weight (g/mol) 154.231
MDL Number MFCD00038027
InChI Key XYHPPOMSLGJAAM-UHFFFAOYSA-N
Synonym 2-(Methylthio)-3-ethylpyrazine
PubChem CID 175366
IUPAC Name 2-ethyl-3-methylsulfanylpyrazine
SMILES CCC1=NC=CN=C1SC

Specifications

Quantity 5 g
Formula Weight 154.23
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Ethyl-3-(methylthio)pyrazine