missing translation for 'onlineSavingsMsg'
Learn More

2-Ethyl-3-methylpyrazine 98.0+%, TCI America™

Catalog No. E03615ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E03615ML 5 mL
E036125ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E03615ML Supplier TCI America Supplier No. E03615ML

Chemical Identifiers

CAS 15707-23-0
Molecular Formula C7H10N2
Molecular Weight (g/mol) 122.17
MDL Number MFCD00006150
InChI Key LNIMMWYNSBZESE-UHFFFAOYSA-N
Synonym pyrazine, 2-ethyl-3-methyl, 3-ethyl-2-methylpyrazine, 2-methyl-3-ethylpyrazine, 2-ethyl-3-methyl pyrazine, pyrazine, ethylmethyl, 2-ethyl-3-methyl-pyrazine, unii-9gf35mk66u, fema no. 3155, pyrazine, 3-ethyl-2-methyl, 2-ethyl-3-methylpyrazine, 9ci
PubChem CID 27457
IUPAC Name 2-ethyl-3-methylpyrazine
SMILES CCC1=NC=CN=C1C

Specifications

Color Yellow
Boiling Point 70°C
Quantity 5 mL
UN Number 1993
Formula Weight 122.17
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-Ethyl-3-methylpyrazine
RTECSNumber UQ3335000
TSCA Yes