missing translation for 'onlineSavingsMsg'
Learn More

2-Ethyl-3-hydroxy-4-pyrone, TCI America™

Catalog No. H055025G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H055025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H055025G Supplier TCI America Supplier No. H055025G

Chemical Identifiers

CAS 11-8-4940
Molecular Formula C7H8O3
Molecular Weight (g/mol) 140.14
MDL Number MFCD00059795
InChI Key YIKYNHJUKRTCJL-UHFFFAOYSA-N
Synonym ethyl maltol, 2-ethyl-3-hydroxy-4h-pyran-4-one, 2-ethyl-3-hydroxy-4-pyrone, veltol plus, 3-hydroxy-2-ethyl-4-pyrone, ethylmaltol, 2-ethylpyromeconic acid, 4h-pyran-4-one, 2-ethyl-3-hydroxy, 3-hydroxy-2-ethyl-1,4-pyrone, 2-ethyl pyromeconic acid
PubChem CID 21059
IUPAC Name 2-ethyl-3-hydroxypyran-4-one
SMILES CCC1=C(C(=O)C=CO1)O

Specifications

Melting Point 92°C
Color White
Quantity 25 g
Formula Weight 140.14
Physical Form Crystalline Powder
Chemical Name or Material 2-Ethyl-3-hydroxy-4-pyrone
EINECSNumber (5)-0678
RTECSNumber UQ0840000
TSCA Yes