missing translation for 'onlineSavingsMsg'
Learn More

2-Ethoxyphenol 99.0+%, TCI America™

Catalog No. E0230500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E0230500G 500 g
E023025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E0230500G Supplier TCI America Supplier No. E0230500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 94-71-3
Molecular Formula C8H10O2
Molecular Weight (g/mol) 138.166
MDL Number MFCD00002187
InChI Key MOEFFSWKSMRFRQ-UHFFFAOYSA-N
Synonym o-ethoxyphenol, phenol, 2-ethoxy, guaethol, guethol, 2-ethyloxyphenol, guaiethol, pyrocatechol monoethyl ether, catechol monoethyl ether, phenol, o-ethoxy, unii-878iw8p9pw
PubChem CID 66755
IUPAC Name 2-ethoxyphenol
SMILES CCOC1=CC=CC=C1O

Specifications

Color White
Boiling Point 216°C
Quantity 500 g
Formula Weight 138.17
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 2-Ethoxyphenol
EINECSNumber (3)-0568
TSCA Yes