missing translation for 'onlineSavingsMsg'
Learn More

2-(Diphenylphosphino)biphenyl 98.0+%, TCI America™

Catalog No. D46881G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D46881G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D46881G Supplier TCI America Supplier No. D46881G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 13885-09-1
Molecular Formula C24H19P
Molecular Weight (g/mol) 338.39
MDL Number MFCD11559063
InChI Key FNCQSSIMHQVKGF-UHFFFAOYSA-N
Synonym 2-diphenylphosphino biphenyl, 2-diphenylphosphino-biphenyl, 1,1'-biphenyl-2-yldiphenylphosphane, biphenyl-2-yl-diphenyl-phosphane, phosphine, 1,1'-biphenyl yldiphenyl, diphenyl 2-phenylphenyl phosphane, 1,1'-biphenyl-2-yldiphenyl-phosphine, ksc494o1b, 2-biphenylyl diphenylphosphine, diphenylphosphino-1,1'-biphenyl
PubChem CID 12976978
IUPAC Name {[1,1'-biphenyl]-2-yl}diphenylphosphane
SMILES C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1C1=CC=CC=C1

Specifications

Melting Point 82°C
Color White
Quantity 1 g
Formula Weight 338.39
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-(Diphenylphosphino)biphenyl