missing translation for 'onlineSavingsMsg'
Learn More

2-(Diphenylphosphino)benzaldehyde, 97%

Catalog No. AAH6130606
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6130606 5 g
AAH6130603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6130606 Supplier Thermo Scientific Chemicals Supplier No. H6130606
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 50777-76-9 | C19H15OP | 290.30 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 50777-76-9
Molecular Formula C19H15OP
Molecular Weight (g/mol) 290.30
MDL Number MFCD00013367
InChI Key DRCPJRZHAJMWOU-UHFFFAOYSA-N
Synonym 2-diphenylphosphino benzaldehyde, 2-diphenylphosphinobenzaldehyde, o-diphenylphosphino benzaldehyde, 2-formylphenyl diphenylphosphine, 2-diphenylphosphanyl benzaldehyde, benzaldehyde, 2-diphenylphosphino, dppbde, zlchem 984, 2-formylphenyldiphenylphosphine
PubChem CID 2754316
IUPAC Name 2-(diphenylphosphanyl)benzaldehyde
SMILES O=CC1=CC=CC=C1P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 112°C to 115°C
Color Yellow
Quantity 5 g
Formula Weight 290.3
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 2-(Diphenylphosphino)benzaldehyde
Hazard Category H302+H332-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only