missing translation for 'onlineSavingsMsg'
Learn More

2-Dimethylaminoethanol, Analytical Standard, ≥99.0% (GC), MilliporeSigma™ Supelco™

Catalog No. 111019442
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019442 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111019442 Supplier Merck Emd Millipore Supplier No. 506321ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 108-01-0
Vapor Pressure 100 mmHg (55°C)
Density 0.886 g/mL (at 20°C)
Physical Form Neat
Linear Formula (CH3)2 NCH2CH2OH
MDL Number MFCD00002846
Melting Point -70°C
Boiling Point 134°C to 136°C
Synonym N,N-Dimethyl-2-hydroxyethylamine; N,N-Dimethylethanolamine; DMEA; Jeffcat DMEA
Solubility Soluble in water
Quantity 1 mL
Formula Weight 89.14
Percent Purity ≥99.0% (GC)
Grade Analytical Standard
Show More Show Less