missing translation for 'onlineSavingsMsg'
Learn More

2-(Dimethylamino)ethyl methacrylate, 97%, stab. with ca 0.2% 4-methoxyphenol

Catalog No. AAL13507AP
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL13507AP 500 mL
AAL13507AE 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL13507AP Supplier Thermo Scientific Chemicals Supplier No. L13507AP
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 2867-47-2 | C8H15NO2 | 157.213 g/mol

2-(Dimethylamino)ethyl methacrylate is used in the synthesis of poly ((2-(Dimethylamino)ethyl methacrylate), block copolymers of 2-(Dimethylamino)ethyl methacrylate and n-butyl methacrylate and amphiphilic AB block copolymers of 2-(Dimethylamino)ethyl methacrylate. It is also used in the preparation of polymer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(Dimethylamino)ethyl methacrylate is used in the synthesis of poly ((2-(Dimethylamino)ethyl methacrylate), block copolymers of 2-(Dimethylamino)ethyl methacrylate and n-butyl methacrylate and amphiphilic AB block copolymers of 2-(Dimethylamino)ethyl methacrylate. It is also used in the preparation of polymer.

Solubility
Soluble in water. (1.000 g/L at 20°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Recommended storage temperature is 2 - 8°C. It is sensitive to moisture and light. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 2867-47-2
Molecular Formula C8H15NO2
Molecular Weight (g/mol) 157.213
MDL Number MFCD00008589
InChI Key JKNCOURZONDCGV-UHFFFAOYSA-N
Synonym 2-dimethylamino ethyl methacrylate, dimethylaminoethyl methacrylate, ageflex fm-1, dmaema, usaf rh-3, 2-propenoic acid, 2-methyl-, 2-dimethylamino ethyl ester, 2-n,n-dimethylamino ethyl methacrylate, n,n-dimethylaminoethyl methacrylate, 2-dimethylaminoethyl methacrylate, n,n-dimethylethanolamine methacrylate
PubChem CID 17869
IUPAC Name 2-(dimethylamino)ethyl 2-methylprop-2-enoate
SMILES CC(=C)C(=O)OCCN(C)C

Specifications

Melting Point ∼-30°C
Density 0.932
Boiling Point 186°C to 188°C
Flash Point 70°C (158°F)
Refractive Index 1.44
Quantity 500 mL
UN Number UN2522
Beilstein 1757048
Solubility Information Soluble in water. (1.000g/L at 20°C).
Formula Weight 157.21
Percent Purity 97%
Chemical Name or Material 2-(Dimethylamino)ethyl methacrylate, stabilized with ≈0.2% 4-methoxyphenol
Show More Show Less
Hazard Category H227-H302-H315-H317-H319-H330
Hazard Statement GHS H Statement
H302-H312-H315-H319-H317-H227
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
Combustible liquid.
Precautionary Statement P210-P233-P240-P241-P242-P243-P260-P264b-P270-P271-P272-P280-P285-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P333+P313-P362-P374-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: 2-DIMETHYLAMINOETHYL METHACRYLATE
EINECSNumber 220-688-8
RTECSNumber OZ4200000
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only