missing translation for 'onlineSavingsMsg'
Learn More

2-(Dimethylamino)ethanol, 99+%

Numéro de catalogue. p-7027111
Change view
Click to view available options
Quantity:
250 mL
1000 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
AAB23616AK 250 mL
AAB23616AU 1000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. AAB23616AK Fournisseur Thermo Scientific Chemicals Code fournisseur B23616AK
Il en reste null

CAS: 108-01-0 | C4H11NO | 89.138 g/mol

2-(Dimethylamino)ethanol is used as corrosion inhibitor, anti-scaling agent, paint additive, coating additive and solids separation agent. It is also used as an intermediate for active pharmaceutical ingredients and dyes. It serves as a curing agent for polyurethanes and epoxy resins. Further, it is used as an additive to boiler water. In addition to this, it is used therapeutically as a CNS stimulant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Miscible with water, alcohol, ether, acetone and benzene.

Notes
Incompatible with strong oxidizers and strong acids.

Identifiants chimiques

CAS 108-01-0
Molecular Formula C4H11NO
Molecular Weight (g/mol) 89.138
MDL Number MFCD00002846
InChI Key UEEJHVSXFDXPFK-UHFFFAOYSA-N
Synonym 2-dimethylamino ethanol, n,n-dimethylethanolamine, deanol, dimethylaminoethanol, dimethylethanolamine, norcholine, dmae, dmea, bimanol, liparon
PubChem CID 7902
ChEBI CHEBI:271436
IUPAC Name 2-(dimethylamino)ethanol
SMILES CN(C)CCO

Spécifications

Melting Point -60°C
Density 0.886
Boiling Point 134°C to 136°C
Flash Point 40°C (104°F)
Odor Ammonia-like
Refractive Index 1.4295
Linear Formula (CH3)2NCH2CH2OH
Quantity 250 mL
UN Number UN2051
Beilstein 1209235
Sensitivity Hygroscopic
Merck Index 14,2843
Solubility Information Miscible with water,alcohol,ether,acetone and benzene.
Formula Weight 89.14
Percent Purity ≥99%
Chemical Name or Material 2-(Dimethylamino)ethanol
Afficher plus de résultats Afficher moins de résultats
Hazard Category H226-H302+H312-H314-H331-H335
Hazard Statement GHS H Statement
H314-H226-H302-H312-H332
Causes severe skin burns and eye damage.
Flammable liquid and vapour.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: 2-DIMETHYLAMINOETHANOL
EINECSNumber 203-542-8
RTECSNumber KK6125000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only