missing translation for 'onlineSavingsMsg'
Learn More

2-(Dimethylamino)-6-propionylnaphthalene 98.0+%, TCI America™

Catalog No. D4678200MG
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D4678200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D4678200MG Supplier TCI America Supplier No. D4678200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 70504-01-7
Molecular Formula C15H17NO
Molecular Weight (g/mol) 227.31
MDL Number MFCD00056615
InChI Key MPPQGYCZBNURDG-UHFFFAOYSA-N
Synonym Prodan
PubChem CID 107729
ChEBI CHEBI:51909
IUPAC Name 1-[6-(dimethylamino)naphthalen-2-yl]propan-1-one
SMILES CCC(=O)C1=CC=C2C=C(C=CC2=C1)N(C)C

Specifications

Melting Point 138°C
Color Yellow
Quantity 200 mg
Formula Weight 227.31
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-(Dimethylamino)-6-propionylnaphthalene