missing translation for 'onlineSavingsMsg'
Learn More

2'-Deoxyinosine-5'-monophosphate disodium salt, 99%, Thermo Scientific Chemicals

Catalog No. AAJ6427103
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6427103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ6427103 Supplier Thermo Scientific Chemicals Supplier No. J6427103

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 14999-52-1 | Molecular Formula: C10H11N4Na2O7P | Molecular Weight: 376.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 14999-52-1
Molecular Formula C10H11N4Na2O7P
Molecular Weight (g/mol) 376.17
MDL Number MFCD00134874
InChI Key VDPLPYMIJCLJEF-DZVPOJRENA-L
Synonym sodium 2r,3s,5r-3-hydroxy-5-6-oxo-1h-purin-9 6h-yl tetrahydrofuran-2-yl methyl phosphate, 2'-deoxyinosine-5'-monophosphate, disodium salt, 2'-deoxyinosine 5'-monophosphate disodium salt, 2'-deoxyinosine-5'-monophosphate disodium salt, disodium dimp, 2'-deoxyinosine-5'-phosphate sodium, disodium 2'-deoxyinosine 5'-phosphate, sodium 2r,3s,5r-3-hydroxy-5-6-hydroxy-9h-purin-9-yl tetrahydrofuran-2-yl methyl phosphate
PubChem CID 56776971
SMILES [Na+].[Na+].O[C@H]1C[C@@H](O[C@@H]1COP([O-])([O-])=O)N1C=NC2=C1N=CNC2=O

Specifications

Color White
Quantity 1 g
IUPAC Name disodium;[(2R,3S,5R)-3-hydroxy-5-(6-oxo-3H-purin-9-yl)oxolan-2-yl]methyl phosphate
Formula Weight 376.17
Percent Purity 99%
Physical Form Powder
Chemical Name or Material 2'-Deoxyinosine-5'-monophosphate disodium salt
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 239-090-3
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only