missing translation for 'onlineSavingsMsg'
Learn More

2'-Deoxyguanosine Hydrate 99.0+%, TCI America™

Catalog No. D00525G
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D00525G 5 g
D00521G 1 g
D005225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D00525G Supplier TCI America Supplier No. D00525G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 961-07-9
Molecular Formula C10H13N5O4
Molecular Weight (g/mol) 267.25
MDL Number MFCD00150760
InChI Key YKBGVTZYEHREMT-DWFKHXNSNA-N
Synonym 2'-deoxyguanosine, deoxyguanosine, guanine deoxyriboside, 2-deoxyguanosine, guanosine, 2'-deoxy, guanine deoxy nucleoside, 2'-deoxy-guanosine, 2;-deoxyguanosine, unii-g9481n71ro, guanine, 9-2-deoxy-beta-d-erythro-pentofuranosyl
PubChem CID 187790
ChEBI CHEBI:17172
IUPAC Name 2-amino-9-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-6,9-dihydro-1H-purin-6-one
SMILES NC1=NC2=C(N=CN2[C@H]2C[C@H](O)[C@@H](CO)O2)C(=O)N1

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 267.25
Percent Purity ≥99.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 2′-Deoxyguanosine Hydrate
EINECSNumber (5)-6830&(8)-0681
RTECSNumber MF8760000
TSCA Yes