missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 2'-Deoxyguanosine-5'-monophosphate disodium salt hydrate, 98%

Catalog No. p-7043040
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6518603 1 g
AAJ6518606 5 g
AAJ6518614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ6518603 Supplier Thermo Scientific Chemicals Supplier No. J6518603

CAS: 146877-98-7 | C10H12N5Na2O7P | 391.19 g/mol

2'-Deoxyguanosine-5'-monophosphate Disodium Salt Hydrate is a reactant involved in analysis of self-assembling in solution and nucleation/growth of G-qudruplexes, Nucleophilic trapping, Reductive alkylation. It is a nucleotide precursor used in DNA synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2′-Deoxyguanosine-5′-monophosphate Disodium Salt Hydrate is a reactant involved in analysis of self-assembling in solution and nucleation/growth of G-qudruplexes, Nucleophilic trapping, Reductive alkylation. It is a nucleotide precursor used in DNA synthesis.

Solubility
Soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 146877-98-7
Molecular Formula C10H12N5Na2O7P
Molecular Weight (g/mol) 391.19
MDL Number MFCD00080331,MFCD00150758
InChI Key CTPAMSRBXKGZCJ-UHFFFAOYNA-L
Synonym 2'-Deoxy-5'-guanylic acid disodium salt; 5'-dGMP disodium salt
PubChem CID 123133822
IUPAC Name [(2R,3S,5R)-5-(2-amino-6-oxo-3H-purin-9-yl)-3-hydroxyoxolan-2-yl]methyl dihydrogen phosphate;sodium
SMILES [Na+].[Na+].NC1=NC(=O)C2=C(N1)N(C=N2)C1CC(O)C(COP([O-])([O-])=O)O1

Specifications

Melting Point >245°C (decomposition)
Color White
Quantity 1 g
Solubility Information Soluble in water.
Formula Weight 391.18 (Anhydrous)
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 2'-Deoxyguanosine-5'-monophosphate disodium salt hydrate
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only