missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals 2-Deoxy-D-galactose, 99%
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spécifications
Spécifications
| CAS Min % | 98.5 |
| CAS Max % | 100.0 |
| Melting Point | 109.0°C to 111.0°C |
| Color | White to Beige |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 98.5% min. (ELSD) (HPLC) |
| Packaging | Glass bottle |
| Quantity | 1 g |
| Specific Rotation | + 57.00 |
| Formula Weight | 164.16 |
| Afficher plus de résultats |
Sécurité et manipulation
| EINECSNumber | 217-765-3 |
RUO – Research Use Only