missing translation for 'onlineSavingsMsg'
Learn More

2'-Deoxy-5-methylcytidine 98.0+%, TCI America™

Catalog No. D36105G
Change view
Click to view available options
Quantity:
100 mg
500 mg
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D36105G 5 g
D3610100MG 100 mg
D3610500MG 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D36105G Supplier TCI America Supplier No. D36105G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 838-07-3
Molecular Formula C10H15N3O4
Molecular Weight (g/mol) 241.247
MDL Number MFCD00006549
InChI Key LUCHPKXVUGJYGU-XLPZGREQSA-N
Synonym 5-methyl-2'-deoxycytidine, 5-methyldeoxycytidine, 2'-deoxy-5-methylcytidine, unii-b200gv71qm, cytidine, 2'-deoxy-5-methyl, 4-amino-1-2r,4s,5r-4-hydroxy-5-hydroxymethyl tetrahydrofuran-2-yl-5-methylpyrimidin-2 1h-one, 5-mdc, 1-2-deoxy-beta-d-ribofuranosyl-5-methylcytosine, 5-methyl-2-deoxycytidine, 4-25-00-03727 beilstein handbook reference
PubChem CID 440055
ChEBI CHEBI:47876
IUPAC Name 4-amino-1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-methylpyrimidin-2-one
SMILES CC1=CN(C(=O)N=C1N)C2CC(C(O2)CO)O

Specifications

Melting Point 193°C
Color White
Quantity 5 g
Formula Weight 241.25
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2′-Deoxy-5-methylcytidine
RTECSNumber HA3860000
TSCA No