missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Decyn-1-ol 95.0+%, TCI America™
$318.97 - $1,194.23
Chemical Identifiers
| CAS | 4117-14-0 |
|---|---|
| Molecular Formula | C10H18O |
| Molecular Weight (g/mol) | 154.253 |
| MDL Number | MFCD00041567 |
| InChI Key | GFLHGTKUCYPXHB-UHFFFAOYSA-N |
| Synonym | 2-decyn-1-ol, 2-decyne-1-ol, n-heptylhydroxymethylacetylene, unii-l4949b9xh9, decynol, pubchem11035, acmc-1arrx, heptyl hydroxymethyl acetylene, 2-decyn-1-ol, |
| PubChem CID | 77763 |
| IUPAC Name | dec-2-yn-1-ol |
| SMILES | CCCCCCCC#CCO |
Chemical Identifiers
| C10H18O | |
| MFCD00041567 | |
| 2-decyn-1-ol, 2-decyne-1-ol, n-heptylhydroxymethylacetylene, unii-l4949b9xh9, decynol, pubchem11035, acmc-1arrx, heptyl hydroxymethyl acetylene, 2-decyn-1-ol, | |
| dec-2-yn-1-ol |
Specifications
| 4117-14-0 | |
| 113°C | |
| MFCD00041567 | |
| GFLHGTKUCYPXHB-UHFFFAOYSA-N | |
| dec-2-yn-1-ol | |
| 77763 | |
| ≥95.0% (GC) | |
| 2-Decyn-1-ol |
| Yellow | |
| C10H18O | |
| 2-decyn-1-ol, 2-decyne-1-ol, n-heptylhydroxymethylacetylene, unii-l4949b9xh9, decynol, pubchem11035, acmc-1arrx, heptyl hydroxymethyl acetylene, 2-decyn-1-ol, | |
| CCCCCCCC#CCO | |
| 154.253 | |
| 154.25 | |
| Liquid |
Safety and Handling
TSCA : No