missing translation for 'onlineSavingsMsg'
Learn More

2-Decyn-1-ol 95.0+%, TCI America™

Catalog No. D12645ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D12645ML 5 mL
D126425ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D12645ML Supplier TCI America Supplier No. D12645ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4117-14-0
Molecular Formula C10H18O
Molecular Weight (g/mol) 154.253
MDL Number MFCD00041567
InChI Key GFLHGTKUCYPXHB-UHFFFAOYSA-N
Synonym 2-decyn-1-ol, 2-decyne-1-ol, n-heptylhydroxymethylacetylene, unii-l4949b9xh9, decynol, pubchem11035, acmc-1arrx, heptyl hydroxymethyl acetylene, 2-decyn-1-ol,
PubChem CID 77763
IUPAC Name dec-2-yn-1-ol
SMILES CCCCCCCC#CCO

Specifications

Color Yellow
Boiling Point 113°C
Quantity 5 mL
Formula Weight 154.25
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Decyn-1-ol