missing translation for 'onlineSavingsMsg'
Learn More

2-Decyn-1-ol 95.0+%, TCI America™

Supplier:  TCI America D126425ML

Quantity 25 mL

 View more versions of this product

Catalog No. D126425ML


Only null left

Description

Description

Specifications

Chemical Identifiers

4117-14-0
154.253
GFLHGTKUCYPXHB-UHFFFAOYSA-N
77763
CCCCCCCC#CCO
C10H18O
MFCD00041567
2-decyn-1-ol, 2-decyne-1-ol, n-heptylhydroxymethylacetylene, unii-l4949b9xh9, decynol, pubchem11035, acmc-1arrx, heptyl hydroxymethyl acetylene, 2-decyn-1-ol,
dec-2-yn-1-ol

Specifications

2-Decyn-1-ol
Yellow
C10H18O
MFCD00041567
GFLHGTKUCYPXHB-UHFFFAOYSA-N
dec-2-yn-1-ol
77763
≥95.0% (GC)
4117-14-0
113°C
25 mL
2-decyn-1-ol, 2-decyne-1-ol, n-heptylhydroxymethylacetylene, unii-l4949b9xh9, decynol, pubchem11035, acmc-1arrx, heptyl hydroxymethyl acetylene, 2-decyn-1-ol,
CCCCCCCC#CCO
154.253
154.25
Liquid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions