accessibility menu, dialog, popup

UserName

H3COH

CAS RN 4117-14-0

IUPAC Name: dec-2-yn-1-ol
Synonyms: dec-2-yn-1-ol
Molecular Weight (g/mol): 154.25
Molecular Formula: C10H18O
InChi Key: GFLHGTKUCYPXHB-UHFFFAOYSA-N
SMILES: CCCCCCCC#CCO
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)