missing translation for 'onlineSavingsMsg'
Learn More

2-Cyclohexylethyl bromide, 98%, Thermo Scientific™

Catalog No. AC392640050
Change view
Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC392640050 5g Glass bottle
AC392640250 25g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC392640050 Supplier Thermo Scientific Chemicals Supplier No. 392640050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 1647-26-3
Molecular Formula C8H15Br
Molecular Weight (g/mol) 191.11
MDL Number MFCD00019398
InChI Key JRQAAYVLPPGEHT-UHFFFAOYSA-N
Synonym 2-bromoethyl cyclohexane, 2-cyclohexylethyl bromide, cyclohexane, 2-bromoethyl, 2-cyclohexylethylbromide, 1-bromo-2-cyclohexylethane, cyclohexylethylbromide, bromoethyl cyclohexane, cyclohexylethyl bromide, acmc-1bzct, 1-bromo-2cyclohexylethane
PubChem CID 15440
IUPAC Name 2-bromoethylcyclohexane
SMILES BrCCC1CCCCC1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Density 1.2210g/mL
Boiling Point 209°C
Flash Point 96°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4890 to 1.4910 (20°C, 589nm)
Linear Formula C6H11(CH2)2Br
Quantity 5g
Specific Gravity 1.221
Formula Weight 191.11
Percent Purity 98%
Chemical Name or Material 2-Cyclohexylethyl bromide
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 216-712-1

RUO – Research Use Only