missing translation for 'onlineSavingsMsg'
Learn More

2-Cyclohexen-1-one 96.0+%, TCI America™

Catalog No. C0707500ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C0707500ML 500 mL
C070725ML 25 mL
C0707100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. C0707500ML Supplier TCI America Supplier No. C0707500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 930-68-7
Molecular Formula C6H8O
Molecular Weight (g/mol) 96.13
MDL Number MFCD00001577
InChI Key FWFSEYBSWVRWGL-UHFFFAOYSA-N
Synonym 2-cyclohexen-1-one, cyclohex-2-enone, 2-cyclohexenone, cyclohexenone, 3-oxocyclohexene, 1-cyclohexen-3-one, cyclohexen-3-one, cyclohexen-1-one, 2-cyclohexenone-1, 2-cyclohexene-1-one
PubChem CID 13594
ChEBI CHEBI:15977
IUPAC Name cyclohex-2-en-1-one
SMILES O=C1CCCC=C1

Specifications

Color Yellow
Boiling Point 171°C
Quantity 500 mL
UN Number 2929
Formula Weight 96.13
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Cyclohexen-1-one
RTECSNumber GW7000000
TSCA Yes