missing translation for 'onlineSavingsMsg'
Learn More

2-Cyclohexen-1-one, 97%

Catalog No. AC111245000
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
500 g
Packaging:
Glass bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC111245000 500 g Glass bottle
AC111240100 10 g Glass bottle
AC111240250 25 g Glass bottle
AC111241000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC111245000 Supplier Thermo Scientific Chemicals Supplier No. 111245000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 930-68-7 | C6H8O | 96.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 930-68-7
Molecular Formula C6H8O
Molecular Weight (g/mol) 96.13
MDL Number MFCD00001577
InChI Key FWFSEYBSWVRWGL-UHFFFAOYSA-N
Synonym 2-cyclohexen-1-one, cyclohex-2-enone, 2-cyclohexenone, cyclohexenone, 3-oxocyclohexene, 1-cyclohexen-3-one, cyclohexen-3-one, cyclohexen-1-one, 2-cyclohexenone-1, 2-cyclohexene-1-one
PubChem CID 13594
ChEBI CHEBI:15977
IUPAC Name cyclohex-2-en-1-one
SMILES O=C1CCCC=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point -53.0°C
Color Yellow
Density 0.9930g/mL
Boiling Point 168.0°C
Flash Point 59°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Refractive Index 1.4875 to 1.4895
Linear Formula C6H8(=O)
Quantity 500 g
Beilstein 07,II,55
Fieser 01,18
Specific Gravity 0.993
Solubility Information Solubility in water: soluble. Other solubilities: soluble in organic solvents
Formula Weight 96.13
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2-Cyclohexen-1-one
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes serious eye irritation.
Fatal in contact with skin.
Fatal if inhaled.
Harmful if swallowed.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF IN EYES: Rinse cautiou
EINECSNumber 213-223-5

RUO – Research Use Only