Learn More
2-Cyanothioacetamide, 98%
CAS: 7357-70-2 | C3H4N2S | 100.14 g/mol
Supplier: Thermo Scientific Chemicals A1543614
Description
2-Cyanothioacetamide was used in the synthesis of 3,4-trans-4-aryl-3-(1-pyridinio)-1,2,3,4-tetrahydropyridine-6-thiolates and bis[6-(2-aryl)-2-thioxo-1,2-dihydropyridine-3-carbonitrile]. 2-Cyanothioacetamide is used as a building block for 2-pyridothiones. It can be used in agrochemical, pharmaceutical and dyestuff field etc,.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Cyanothioacetamide was used in the synthesis of 3,4-trans-4-aryl-3-(1-pyridinio)-1,2,3,4-tetrahydropyridine-6-thiolates and bis[6-(2-aryl)-2-thioxo-1,2-dihydropyridine-3-carbonitrile]. 2-Cyanothioacetamide is used as a building block for 2-pyridothiones. It can be used in agrochemical, pharmaceutical and dyestuff field etc,.
Solubility
Does not mix well with water.
Notes
Light Sensitive. Store in the dark.
Specifications
| 2-Cyanothioacetamide | |
| 114°C to 120°C | |
| MFCD00010025 | |
| UN3439 | |
| Light sensitive | |
| Does not mix well with water. | |
| NC(=S)CC#N | |
| 100.14 | |
| 100.14 |
| 7357-70-2 | |
| C3H4N2S | |
| 25 g | |
| 1699934 | |
| 2-cyanothioacetamide, cyanothioacetamide, ethanethioamide, 2-cyano, cyano thioacetamide, 2-cyano-thioacetamide, 3-amino-3-thioxopropanenitrile, cyanthioacetamide, cyanoacetothioamide, 2-cyanothioactamide, alpha-cyanothioacetamide | |
| BHPYMZQTCPRLNR-UHFFFAOYSA-N | |
| 2-cyanoethanethioamide | |
| 1416277 | |
| 98% |
Chemical Identifiers
| 7357-70-2 | |
| 100.14 | |
| BHPYMZQTCPRLNR-UHFFFAOYSA-N | |
| 1416277 | |
| NC(=S)CC#N |
| C3H4N2S | |
| MFCD00010025 | |
| 2-cyanothioacetamide, cyanothioacetamide, ethanethioamide, 2-cyano, cyano thioacetamide, 2-cyano-thioacetamide, 3-amino-3-thioxopropanenitrile, cyanthioacetamide, cyanoacetothioamide, 2-cyanothioactamide, alpha-cyanothioacetamide | |
| 2-cyanoethanethioamide |
Safety and Handling
GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only