accessibility menu, dialog, popup

UserName

CAS RN 7357-70-2

IUPAC Name: 2-cyanoethanethioamide
Synonyms: 2-cyanoethanethioamide
Molecular Weight (g/mol): 100.14
Molecular Formula: C3H4N2S
InChi Key: BHPYMZQTCPRLNR-UHFFFAOYSA-N
SMILES: NC(=S)CC#N
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (5)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)