missing translation for 'onlineSavingsMsg'
Learn More

2-Cyanothioacetamide, 98%

Catalog No. p-7022588
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1543606 5 g
AAA1543614 25 g
AAA1543622 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1543606 Supplier Thermo Scientific Chemicals Supplier No. A1543606
Only null left

CAS: 7357-70-2 | C3H4N2S | 100.14 g/mol

2-Cyanothioacetamide was used in the synthesis of 3,4-trans-4-aryl-3-(1-pyridinio)-1,2,3,4-tetrahydropyridine-6-thiolates and bis[6-(2-aryl)-2-thioxo-1,2-dihydropyridine-3-carbonitrile]. 2-Cyanothioacetamide is used as a building block for 2-pyridothiones. It can be used in agrochemical, pharmaceutical and dyestuff field etc,.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Cyanothioacetamide was used in the synthesis of 3,4-trans-4-aryl-3-(1-pyridinio)-1,2,3,4-tetrahydropyridine-6-thiolates and bis[6-(2-aryl)-2-thioxo-1,2-dihydropyridine-3-carbonitrile]. 2-Cyanothioacetamide is used as a building block for 2-pyridothiones. It can be used in agrochemical, pharmaceutical and dyestuff field etc,.

Solubility
Does not mix well with water.

Notes
Light Sensitive. Store in the dark.

Chemical Identifiers

CAS 7357-70-2
Molecular Formula C3H4N2S
Molecular Weight (g/mol) 100.14
MDL Number MFCD00010025
InChI Key BHPYMZQTCPRLNR-UHFFFAOYSA-N
Synonym 2-cyanothioacetamide, cyanothioacetamide, ethanethioamide, 2-cyano, cyano thioacetamide, 2-cyano-thioacetamide, 3-amino-3-thioxopropanenitrile, cyanthioacetamide, cyanoacetothioamide, 2-cyanothioactamide, alpha-cyanothioacetamide
PubChem CID 1416277
IUPAC Name 2-cyanoethanethioamide
SMILES NC(=S)CC#N

Specifications

Melting Point 114°C to 120°C
Quantity 5 g
UN Number UN3439
Beilstein 1699934
Sensitivity Light sensitive
Solubility Information Does not mix well with water.
Formula Weight 100.14
Percent Purity 98%
Chemical Name or Material 2-Cyanothioacetamide
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only