missing translation for 'onlineSavingsMsg'
Learn More

2-Cyanophenol, 99%

Catalog No. AC161410050
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC161410050 5 g Glass bottle
AC161410250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC161410050 Supplier Thermo Scientific Chemicals Supplier No. 161410050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 611-20-1 | C7H5NO | 119.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 611-20-1
Molecular Formula C7H5NO
Molecular Weight (g/mol) 119.12
MDL Number MFCD00002145
InChI Key CHZCERSEMVWNHL-UHFFFAOYSA-N
Synonym 2-cyanophenol, o-cyanophenol, salicylonitrile, o-hydroxybenzonitrile, benzonitrile, 2-hydroxy, salicylnitrile, benzonitrile, hydroxy, benzonitrile, o-hydroxy, o-hydoxybenzonitrile, 2-hydroxy-benzonitrile
PubChem CID 11907
IUPAC Name 2-hydroxybenzonitrile
SMILES C1=CC=C(C(=C1)C#N)O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 95.0°C to 98.0°C
Color White to Brown
Boiling Point 149.0°C (14.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula NCC6H4OH
Quantity 5 g
Beilstein 10, 96
Solubility Information Solubility in water: soluble
Formula Weight 119.12
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 2-Cyanophenol, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if swallowed.
May cause an allergic skin reaction.
Causes serious eye damage.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Con
EINECSNumber 210-259-3
TSCA TSCA

RUO – Research Use Only