missing translation for 'onlineSavingsMsg'
Learn More

2-Cyano-6-methoxybenzothiazole, 99%, Thermo Scientific™

Catalog No. AC225362500
Change view
Click to view available options
250mg; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC225362500 250mg; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC225362500 Supplier Thermo Scientific Chemicals Supplier No. 225362500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 943-03-3
CAS Min % 98.5
CAS Max % 100.0
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Molecular Formula C9H6N2OS
Quantity 250mg
MDL Number MFCD00010537
Synonym 2-cyano-6-methoxybenzothiazole, 6-methoxybenzo d thiazole-2-carbonitrile, 6-methoxy-2-benzothiazolecarbonitrile, 6-methoxybenzothiazole-2-carbonitrile, 6-methoxy-benzothiazole-2-carbonitrile, 6-methoxy-2-cyano-benzothiazole, 2-benzothiazolecarbonitrile, 6-methoxy, pubchem9766, acmc-209rr8, ksc486m5p
InChI Key DEWDWBYQOFXKIH-UHFFFAOYSA-N
SMILES COC1=CC2=C(C=C1)N=C(S2)C#N
IUPAC Name 6-methoxy-1,3-benzothiazole-2-carbonitrile
Molecular Weight (g/mol) 190.23
PubChem CID 342109
Formula Weight 190.23
Percent Purity 99%
Chemical Name or Material 2-Cyano-6-methoxybenzothiazole
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if inhaled.
Harmful in contact with skin.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only