missing translation for 'onlineSavingsMsg'
Learn More

2-Cyano-3-methylpyridine 98.0+%, TCI America™

Catalog No. C14105G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C14105G 5 g
C141025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
Catalog No. C14105G missing translation for 'mfr' TCI America missing translation for 'supplierNo' C14105G

Chemical Identifiers

CAS 20970-75-6
Molecular Formula C7H6N2
Molecular Weight (g/mol) 118.139
MDL Number MFCD00190585
InChI Key WBXZCDIZXWDPBL-UHFFFAOYSA-N
Synonym 2-cyano-3-methylpyridine, 3-methylpicolinonitrile, 2-cyano-3-methyl pyridine, 3-methyl-2-pyridinecarbonitrile, pyridinecarbonitrile, methyl, 3-methyl-pyridine-2-carbonitrile, 2-cyano-3-methyl-pyridine, 2-pyridinecarbonitrile, 3-methyl, 2-cyano-3-methylpyridine 2-cyano-3-picoline, 2-cyano-3-picoline
PubChem CID 819928
IUPAC Name 3-methylpyridine-2-carbonitrile
SMILES CC1=C(N=CC=C1)C#N

Specifications

Melting Point 86°C
Color White-Yellow
Boiling Point 142°C
Quantity 5 g
UN Number 3439
Formula Weight 118.14
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Cyano-3-methylpyridine