missing translation for 'onlineSavingsMsg'
Learn More

2-Cyano-3-fluoropyridine, 97+%, Thermo Scientific Chemicals

Catalog No. AAH2576706
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2576706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH2576706 Supplier Thermo Scientific Chemicals Supplier No. H2576706
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 97509-75-6 | Molecular Formula: C6H3FN2 | Molecular Weight: 122.102 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 97509-75-6
Molecular Formula C6H3FN2
Molecular Weight (g/mol) 122.102
MDL Number MFCD06797501
InChI Key VZFPSCNTFBJZHB-UHFFFAOYSA-N
Synonym 2-cyano-3-fluoropyridine, 3-fluoropicolinonitrile, 3-fluoro-2-pyridinecarbonitrile, 3-fluoro-2-cyanopyridine, 3-fluoro-2-pyridinenitrile, 3-fluoro-pyridine-2-carbonitrile, 2-pyridinecarbonitrile, 3-fluoro, fluorocyanopyridine, pubchem16434, acmc-209s8i
PubChem CID 7060408
IUPAC Name 3-fluoropyridine-2-carbonitrile
SMILES C1=CC(=C(N=C1)C#N)F

Specifications

Melting Point 27°C to 30°C
Quantity 5 g
Formula Weight 122.1
Percent Purity ≥97%
Chemical Name or Material 2-Cyano-3-fluoropyridine
Hazard Category H302+H312+H332
Hazard Statement GHS H Statement
H302-H315-H319
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only