missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorothioxanthone 98,0+%, TCI America™

Numéro de catalogue. p-7124341
Modifier l'affichage
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
C02925G 5 g
C029225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. C02925G Fournisseur TCI America Code fournisseur C02925G

Identifiants chimiques

CAS 86-39-5
Molecular Formula C13H7ClOS
Molecular Weight (g/mol) 246.71
MDL Number MFCD00005067
InChI Key ZCDADJXRUCOCJE-UHFFFAOYSA-N
Synonym 2-chlorothioxanthone, 2-chloro-9h-thioxanthen-9-one, 9h-thioxanthen-9-one, 2-chloro, unii-rgs87t004b, 2-chlorothiaxanthone, 2-chlorothioxanthene-9-one, sandoray 1050, 2-chloro-thioxanthone, pubchem10692, 2-chloro-9-thioxanthone
PubChem CID 618848
IUPAC Name 2-chloro-9H-thioxanthene-9-one
SMILES ClC1=CC=C2SC3=CC=CC=C3C(=O)C2=C1

Spécifications

Melting Point 152°C
Quantity 5 g
UN Number 1325
Formula Weight 246.71
Percent Purity ≥98.0% (GC)
Physical Form Poudre cristalline
Chemical Name or Material 2-Chlorothioxanthone
EINECSNumber (5)-5537
TSCA Yes