missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorothioxanthone, British Pharmacopoeia (BP) Reference Standard, MilliporeSigma™ Supelco™

Catalog No. 111022905
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111022905 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111022905 Supplier Merck Emd Millipore Supplier No. BP559

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets, has been developed and issued under the Authority of the issuing Pharmacopoeia.

Specifications

CAS 86-39-5
Melting Point 153°C to 154°C (literature)
Molecular Formula C13H7ClOS
Linear Formula C13H7ClOS
Quantity 25 mg
MDL Number MFCD00005067
Synonym 2-Chlorothioxanthen-9-one
Formula Weight 246.71
Grade BP