missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorothioxanthone, 99%

Catalog No. AAA1813122
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1813122 100 g
AAA1813106 5 g
AAA1813114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1813122 Supplier Thermo Scientific Chemicals Supplier No. A1813122
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 86-39-5 | C13H7ClOS | 246.71 g/mol

2-Chlorothioxanthone is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chlorothioxanthone is used as pharmaceutical intermediate.

Solubility
Partly miscible in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 86-39-5
Molecular Formula C13H7ClOS
Molecular Weight (g/mol) 246.71
MDL Number MFCD00005067
InChI Key ZCDADJXRUCOCJE-UHFFFAOYSA-N
Synonym 2-chlorothioxanthone, 2-chloro-9h-thioxanthen-9-one, 9h-thioxanthen-9-one, 2-chloro, unii-rgs87t004b, 2-chlorothiaxanthone, 2-chlorothioxanthene-9-one, sandoray 1050, 2-chloro-thioxanthone, pubchem10692, 2-chloro-9-thioxanthone
PubChem CID 618848
IUPAC Name 2-chloro-9H-thioxanthen-9-one
SMILES ClC1=CC=C2SC3=CC=CC=C3C(=O)C2=C1

Specifications

Melting Point 151°C to 154°C
Flash Point 91°C (196°F)
Quantity 100 g
Solubility Information Partly miscible in water.
Formula Weight 246.72
Percent Purity 99%
Chemical Name or Material 2-Chlorothioxanthone
EINECSNumber 201-667-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only