missing translation for 'onlineSavingsMsg'
Learn More

2-Chloropropionic Acid 98.0+%, TCI America™

Catalog No. C0275500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C0275500G 500 g
C027525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C0275500G Supplier TCI America Supplier No. C0275500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 598-78-7
Molecular Formula C3H5ClO2
Molecular Weight (g/mol) 108.52
MDL Number MFCD00004224
InChI Key GAWAYYRQGQZKCR-UHFFFAOYNA-N
Synonym 2-chloropropionic acid, propanoic acid, 2-chloro, propionic acid, 2-chloro, alpha-chloropropionic acid, alpha-monochloropropionic acid, propionic acid, alpha-chloro, .alpha.-chloropropionic acid, propionic acid, .alpha.-chloro, chloropropionic acid, propanoic acid, chloro
PubChem CID 11734
IUPAC Name 2-chloropropanoic acid
SMILES CC(Cl)C(O)=O

Specifications

Melting Point -12°C
Color Yellow
Boiling Point 186°C
Quantity 500 g
UN Number 2511
Formula Weight 108.52
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-Chloropropionic Acid
EINECSNumber (2)-1157
RTECSNumber UE8575000
TSCA Yes