missing translation for 'onlineSavingsMsg'
Learn More

(+/-)-2-Chloropropionic acid, 94%

Catalog No. AAA1286022
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1286022 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1286022 Supplier Thermo Scientific Chemicals Supplier No. A1286022
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 598-78-7 | C3H5ClO2 | 108.52 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

 

Chemical Identifiers

CAS 598-78-7
Molecular Formula C3H5ClO2
Molecular Weight (g/mol) 108.52
MDL Number MFCD00004224
InChI Key GAWAYYRQGQZKCR-UHFFFAOYNA-N
Synonym 2-chloropropionic acid, propanoic acid, 2-chloro, propionic acid, 2-chloro, alpha-chloropropionic acid, alpha-monochloropropionic acid, propionic acid, alpha-chloro, .alpha.-chloropropionic acid, propionic acid, .alpha.-chloro, chloropropionic acid, propanoic acid, chloro
PubChem CID 11734
IUPAC Name 2-chloropropanoic acid
SMILES CC(Cl)C(O)=O

Specifications

Melting Point -13°C
Density 1.262
Boiling Point 78°C to 80°C (11 mmHg)
Flash Point 101°C (213°F)
Refractive Index 1.435
Quantity 100 g
UN Number UN2511
Beilstein 1720259
Formula Weight 108.53
Percent Purity 94%
Chemical Name or Material (+/-)-2-Chloropropionic acid
Show More Show Less
Hazard Category H302-H314
Hazard Statement GHS H Statement
H314-H302
Causes severe skin burns and eye damage.
Harmful if swallowed.
Precautionary Statement P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: 2-CHLOROPROPIONIC ACID
EINECSNumber 209-952-3
RTECSNumber UE8575000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only