missing translation for 'onlineSavingsMsg'
Learn More

2-chlorophenylacetyl Chloride, 97%

Catalog No. AC395080250
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC395080250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC395080250 Supplier Thermo Scientific Chemicals Supplier No. 395080250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 51512-09-5 | C8H6Cl2O | 189.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 51512-09-5
Molecular Formula C8H6Cl2O
Molecular Weight (g/mol) 189.04
MDL Number MFCD00130223
InChI Key WIHSAOYVGKVRJX-UHFFFAOYSA-N
Synonym 2-chlorophenylacetyl chloride, 2-2-chlorophenyl acetyl chloride, acetyl chloride, 2-2-chlorophenyl, o-chlorophenylacetyl chloride, benzeneacetyl chloride, 2-chloro, 2-chlorophenylacetyl chloride stabilized with copper chip, acmc-1auqq, o-chlor-phenylacetylchlorid, o-chloro-phenylacetyl chloride, 2-chloro-phenylacetyl chloride
PubChem CID 182669
IUPAC Name 2-(2-chlorophenyl)acetyl chloride
SMILES ClC(=O)CC1=CC=CC=C1Cl

Specifications

Color Yellow
CAS Min % 96.0
CAS Max % 100.0
Density 1.3100g/mL
Quantity 25 g
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Liquid
Refractive Index 1.5480 to 1.55
Specific Gravity 1.31
Assay Percent Range 97%
Linear Formula ClC6H4CH2COCl
Formula Weight 189.04
Percent Purity 97%
Chemical Name or Material 2-chlorophenylacetyl chloride
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes serious eye damage.
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only