missing translation for 'onlineSavingsMsg'
Learn More

2-Chloronicotinic acid, 99%

Catalog No. AC404860500
Change view
Click to view available options
Quantity:
50 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC404860500 50 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC404860500 Supplier Thermo Scientific Chemicals Supplier No. 404860500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 2942-59-8 | C6H4ClNO2 | 157.56 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2942-59-8
Molecular Formula C6H4ClNO2
Molecular Weight (g/mol) 157.56
MDL Number MFCD00006236
InChI Key IBRSSZOHCGUTHI-UHFFFAOYSA-N
Synonym 2-chloronicotinic acid, 2-chloro-3-pyridinecarboxylic acid, 3-pyridinecarboxylic acid, 2-chloro, 2-chloro-nicotinic acid, 2-chloro nicotinic acid, chloronicotinic acid, nicotinic acid, 2-chloro, 2-chloronicotinicacid, 2-chloroisonicotinicacid, 2chloronicotinic acid
PubChem CID 76258
IUPAC Name 2-chloropyridine-3-carboxylic acid
SMILES C1=CC(=C(N=C1)Cl)C(=O)O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 176°C to 178°C
Color White to Cream
Loss on Drying 0.3% max. (105°C, 3 hrs)
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Linear Formula Cl(C5H3N)COOH
Quantity 50 g
Beilstein 22, II, 35
Formula Weight 157.56
Percent Purity 99%
Physical Form Powder
Chemical Name or Material 2-Chloronicotinic acid, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF
EINECSNumber 220-937-

RUO – Research Use Only