missing translation for 'onlineSavingsMsg'
Learn More

2-Chloronaphtalène 98,0+%, TCI America™

Numéro de catalogue. C02131G
Modifier l'affichage
Click to view available options
Quantity:
1 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
C02131G 1 g
C021325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. C02131G Fournisseur TCI America Code fournisseur C02131G

Identifiants chimiques

CAS 91-58-7
Molecular Formula C10H7Cl
Molecular Weight (g/mol) 162.616
MDL Number MFCD00035731
InChI Key CGYGETOMCSJHJU-UHFFFAOYSA-N
Synonym naphthalene, 2-chloro, 2-chloro naphthalene, beta-chloronaphthalene, halowax, 2-chlornaftalen, rcra waste number u047, 2-chloro-naphthalene, .beta.-chloronaphthalene, 2-chlornaftalen czech, unii-49o81u3iti
PubChem CID 7056
IUPAC Name 2-chloronaphtalène
SMILES C1=CC=C2C=C(C=CC2=C1)Cl

Spécifications

Melting Point 59°C
Color Blanc
Boiling Point 256°C
Quantity 1 g
Formula Weight 162.62
Percent Purity ≥98.0% (GC)
Physical Form Poudre cristalline
Chemical Name or Material 2-Chloronaphthalene
EINECSNumber (4)-0316
RTECSNumber QJ2275000
TSCA Yes