missing translation for 'onlineSavingsMsg'
Learn More

2-(Chloromethyl)-1,2-epoxypropane 98.0+%, TCI America™

Catalog No. C25845G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C25845G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C25845G Supplier TCI America Supplier No. C25845G

Chemical Identifiers

CAS 598-09-4
Molecular Formula C4H7ClO
Molecular Weight (g/mol) 106.549
MDL Number MFCD00052487
InChI Key VVHFXJOCUKBZFS-UHFFFAOYSA-N
Synonym 2-chloromethyl-2-methyloxirane, oxirane, 2-chloromethyl-2-methyl, 2-chloromethyl-2-methyl-oxirane, 2-chloromethyl-1,2-epoxypropane, methyl epichlorohydrin, 2-methylepichlorohydrin, acmc-209mfx, beta-methylepichlorohydrin, ksc493o4b
PubChem CID 95220
IUPAC Name 2-(chloromethyl)-2-methyloxirane
SMILES CC1(CO1)CCl

Specifications

Color Colorless
Boiling Point 122°C
Quantity 5 g
UN Number 2920
Formula Weight 106.55
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-(Chloromethyl)-1,2-epoxypropane
EINECSNumber (2)-2403
TSCA No