missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chloroisonicotinaldehyde, 97%, Thermo Scientific™
Spécifications
Spécifications
| Nom ou substance chimique | 2-Chloroisonicotinaldehyde |
| Point d’éclair | >110°C |
| Spectre infrarouge | Authentic |
| Fourchette de pourcentages d’essai | 96% min. (GC) |
| Emballage | Glass Bottle |
| Quantité | 1g |
| Poids de formule | 141.56 |
| Pourcentage de pureté | 97% |
Sécurité et manipulation
| Hazard Category | GHS Signal Word: Warning |
| Hazard Statement | GHS H Statement Harmful if inhaled. Harmful if swallowed. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. Harmful in contact with skin. |
| Precautionary Statement | GHS P Statement IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses,if present and easy to do. Continue rinsing. IF ON SKIN: Wash with plenty of soap and water. Avoid breathing dust/fume/gas/mist/vapo |
RUO – Research Use Only