missing translation for 'onlineSavingsMsg'
Learn More

2-Chloroheptane 97.0+%, TCI America™

Catalog No. p-7124644
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C08525ML 5 mL
C085225ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C08525ML Supplier TCI America Supplier No. C08525ML

Chemical Identifiers

CAS 1001-89-4
Molecular Formula C7H15Cl
Molecular Weight (g/mol) 134.65
MDL Number MFCD00039353
InChI Key PTSLUOSUHFGQHV-UHFFFAOYNA-N
Synonym 2-Heptyl Chloride
PubChem CID 34691
IUPAC Name 2-chloroheptane
SMILES CCCCCC(C)Cl

Specifications

Color Colorless
Boiling Point 146°C
Quantity 5 mL
UN Number 1993
Formula Weight 134.65
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Chloroheptane