missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorobenzylamine 98.0+%, TCI America™

Catalog No. C106525G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C106525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C106525G Supplier TCI America Supplier No. C106525G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 89-97-4
Molecular Formula C7H8ClN
Molecular Weight (g/mol) 141.598
MDL Number MFCD00008108
InChI Key KDDNKZCVYQDGKE-UHFFFAOYSA-N
Synonym 2-chlorobenzylamine, 2-chlorophenyl methanamine, o-chlorobenzylamine, benzenemethanamine, 2-chloro, o-chlorobenzyl amine, 2-chlorobenzenemethanamine, 2-chloro-benzylamine, benzylamine, o-chloro, 2-chlorophenyl methylamine, unii-l790j4y7a3
PubChem CID 66648
IUPAC Name (2-chlorophenyl)methanamine
SMILES C1=CC=C(C(=C1)CN)Cl

Specifications

Color Yellow
Boiling Point 219°C
Quantity 25 g
UN Number 2735
Formula Weight 141.60
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Chlorobenzylamine
EINECSNumber (3)-4353
TSCA Yes