missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorobenzyl Bromide 98.0+%, TCI America™

Catalog No. C14435G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C14435G 5 g
C144325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C14435G Supplier TCI America Supplier No. C14435G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 611-17-6
Molecular Formula C7H6BrCl
Molecular Weight (g/mol) 205.479
MDL Number MFCD00000566
InChI Key PURSZYWBIQIANP-UHFFFAOYSA-N
Synonym 2-chlorobenzyl bromide, 1-bromomethyl-2-chlorobenzene, o-chlorobenzyl bromide, 2-chlorobenzylbromide, benzene, 1-bromomethyl-2-chloro, alpha-bromo-2-chlorotoluene, unii-cev23h8uv4, 2chlorobenzyl bromide, alpha-bromo-o-chlorotoluene, cev23h8uv4
PubChem CID 11905
IUPAC Name 1-(bromomethyl)-2-chlorobenzene
SMILES C1=CC=C(C(=C1)CBr)Cl

Specifications

Color Yellow
Boiling Point 130°C
Quantity 5 g
UN Number 3265
Formula Weight 205.48
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Chlorobenzyl Bromide