missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chlorobenzoyl chloride, 98%, Thermo Scientific™
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Spécifications
Spécifications
| Nom ou substance chimique | 2-Chlorobenzoyl chloride |
| Densité | 1.3800g/mL |
| Point d’ébullition | 238.0°C |
| Point d’éclair | 124°C |
| Spectre infrarouge | Authentic |
| Fourchette de pourcentages d’essai | 97.5% min. (GC) |
| Emballage | Glass Bottle |
| Indice de réfraction | 1.5708 to 1.5728 |
| Formule linéaire | ClC6H4COCl |
| Quantité | 1L |
| Afficher plus de résultats |
Sécurité et manipulation
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement Causes severe skin burns and eye damage. May be corrosive to metals. |
| Precautionary Statement | GHS P Statement IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses,if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. Wear protective gloves/protective |
| EINECSNumber | 210-194- |
RUO – Research Use Only