missing translation for 'onlineSavingsMsg'
Learn More

2-Chloroaniline Hydrochloride 98.0+%, TCI America™

Catalog No. C011425G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C011425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C011425G Supplier TCI America Supplier No. C011425G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 137-04-2
Molecular Formula C6H7Cl2N
Molecular Weight (g/mol) 164.03
MDL Number MFCD00053282
InChI Key DRGIDRZFKRLQTE-UHFFFAOYSA-N
Synonym 2-chloroaniline hydrochloride, 2-chloroanilinium chloride, benzenamine, 2-chloro-, hydrochloride, benzenamine, 2-chloro-, hydrochloride 1:1, 2-chlorophenylamine, chloride, acmc-209cai, o-chloroaniline hydrochloride, 95-51-2 parent, 2-chloranylaniline hydrochloride, benzenamine,2-chloro-,hydrochloride 1:1
PubChem CID 67298
IUPAC Name hydrogen 2-chloroaniline chloride
SMILES [H+].[Cl-].NC1=CC=CC=C1Cl

Specifications

Color White-Yellow
Quantity 25 g
UN Number 2018
Formula Weight 164.03
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloroaniline Hydrochloride
EINECSNumber (3)-0194
TSCA Yes