missing translation for 'onlineSavingsMsg'
Learn More

2-Chloroaniline hydrochloride, 97%

Catalog No. AAL1348322
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1348322 100 g
AAL1348314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1348322 Supplier Thermo Scientific Chemicals Supplier No. L1348322

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 137-04-2 | C6H7Cl2N | 164.03 g/mol

2-Chloroaniline hydrochloride, is used as ice dye basement, used for towels, colored yarn and cloth of cotton, viscose fabric dyeing and printing of chromogenic agent. It is also used as an intermediate in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chloroaniline hydrochloride, is used as ice dye basement, used for towels, colored yarn and cloth of cotton, viscose fabric dyeing and printing of chromogenic agent. It is also used as an intermediate in organic synthesis.

Solubility
It is soluble in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.

Chemical Identifiers

CAS 137-04-2
Molecular Formula C6H7Cl2N
Molecular Weight (g/mol) 164.03
MDL Number MFCD00053282
InChI Key DRGIDRZFKRLQTE-UHFFFAOYSA-N
Synonym 2-chloroaniline hydrochloride, 2-chloroanilinium chloride, benzenamine, 2-chloro-, hydrochloride, benzenamine, 2-chloro-, hydrochloride 1:1, 2-chlorophenylamine, chloride, acmc-209cai, o-chloroaniline hydrochloride, 95-51-2 parent, 2-chloranylaniline hydrochloride, benzenamine,2-chloro-,hydrochloride 1:1
PubChem CID 67298
IUPAC Name 2-chloroaniline;hydrochloride
SMILES [H+].[Cl-].NC1=CC=CC=C1Cl

Specifications

Melting Point ∼220°C (sublimation)
Odor Characteristic
Quantity 100 g
UN Number UN2018
Beilstein 3624280
Solubility Information It is soluble in water.
Formula Weight 164.03
Percent Purity 97%
Chemical Name or Material 2-Chloroaniline hydrochloride
Hazard Category H301+H311+H331-H373
Hazard Statement GHS H Statement
H301-H311-H331-H373
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Precautionary Statement P260-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P311-P312-P314-P330-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: CHLOROANILINES, SOLID
EINECSNumber 205-274-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only