missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chloroadenosine
Description
2-Chloroadenosine is used as a selective A1-adenosine receptor agonist, and also Induces apoptosis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Chloroadenosine is used as a selective A1-adenosine receptor agonist, and also Induces apoptosis.
Solubility
Soluble in water.
Notes
protect from heat. Store away from oxidizing agents.
2-Chloroadenosine is used as a selective A1-adenosine receptor agonist, and also Induces apoptosis.
Solubility
Soluble in water.
Notes
protect from heat. Store away from oxidizing agents.
Spécifications
Spécifications
| Color | White |
| Quantity | 250 mg |
| Beilstein | 43957 |
| Solubility Information | Soluble in water. |
| Formula Weight | 301.69 |
| Physical Form | Powder |
| Chemical Name or Material | 2-Chloroadenosine |
Sécurité et manipulation
| EINECSNumber | 205-678-3 |
| TSCA | No |
| Recommended Storage | Store at -20°C |
RUO – Research Use Only