missing translation for 'onlineSavingsMsg'
Learn More

2-chloroadénosine hydrate 98,0+%, TCI America™

Numéro de catalogue. C2192100MG
Modifier l'affichage
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
C2192100MG 100 mg
C21921G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. C2192100MG Fournisseur TCI America Code fournisseur C2192100MG

Identifiants chimiques

CAS 146-77-0
Molecular Formula C10H12ClN5O4
Molecular Weight (g/mol) 301.69
MDL Number MFCD00005734
InChI Key BIXYYZIIJIXVFW-SBTUYZBDNA-N
Synonym 2-chloroadenosine, 6-amino-2-chloropurine riboside, adenosine, 2-chloro, cl-ado, 2-chloroadenosinehemihydrate, unii-7w7zug45g8, 2-chloro adenosine, 2cl-ado, cado, 2 clado
PubChem CID 8974
IUPAC Name (2R,3R,4S,5R)-2-(6-amino-2-chloro-9H-purine-9-yl)-5-(hydroxyméthyl)oxolane-3,4-diol
SMILES NC1=C2N=CN([C@@H]3O[C@H](CO)[C@@H](O)[C@H]3O)C2=NC(Cl)=N1

Spécifications

Melting Point 162°C
Color Blanc-jaune
Quantity 100 mg
Formula Weight 301.69
Percent Purity ≥98.0% (HPLC,T)
Physical Form Poudre cristalline
Chemical Name or Material 2-Chloroadenosine Hydrate
RTECSNumber AU7357550
TSCA No