missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-p-xylene 98.0+%, TCI America™

Catalog No. C0313500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C0313500G 500 g
C031325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C0313500G Supplier TCI America Supplier No. C0313500G

Chemical Identifiers

CAS 95-72-7
Molecular Formula C8H9Cl
Molecular Weight (g/mol) 140.61
MDL Number MFCD00000564
InChI Key KZNRNQGTVRTDPN-UHFFFAOYSA-N
Synonym 2-chloro-p-xylene, 1-chloro-2,5-dimethylbenzene, 2,5-dimethylchlorobenzene, benzene, 2-chloro-1,4-dimethyl, chloro-p-xylene, p-xylene, 2-chloro, 2-chloro-1,4-dimethyl-benzene, unii-3y9k758wlv, chloro-p-xylene van, 2-chlor-p-xylol
PubChem CID 32883
IUPAC Name 2-chloro-1,4-dimethylbenzene
SMILES CC1=CC(=C(C=C1)C)Cl

Specifications

Color Colorless
Quantity 500 g
Physical Form Clear Liquid at 20°C
UN Number 1993
Boiling Point 186°C
Formula Weight 140.61
Percent Purity ≥98.0% (GC)
Chemical Name or Material 2-Chloro-p-xylene
EINECSNumber (3)-0055
TSCA Yes