missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-6-methylpyrazine 98.0+%, TCI America™

Catalog No. C27775G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C27775G 5 g
C27771G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C27775G Supplier TCI America Supplier No. C27775G
Only null left

Chemical Identifiers

CAS 38557-71-0
Molecular Formula C5H5ClN2
Molecular Weight (g/mol) 128.56
MDL Number MFCD00055032
InChI Key CKUVSPQGYLELRG-UHFFFAOYSA-N
Synonym 2-chloro-6-methyl-pyrazine, pyrazine, 2-chloro-6-methyl, 6-chloro-2-methylpyrazine, 2-chloro-6-methyl-1,4-diazine, 2-methyl-6-chloropyrazine, pubchem18034, acmc-1afqe, 2-chloro-6 methylpyrazine, ksc497m1j
PubChem CID 7170095
IUPAC Name 2-chloro-6-methylpyrazine
SMILES CC1=CN=CC(Cl)=N1

Specifications

Melting Point 52°C
Color White
Boiling Point 83°C
Quantity 5 g
Formula Weight 128.56
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloro-6-methylpyrazine