missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-6-fluorobenzoyl Chloride 98.0+%, TCI America™

Catalog No. C22295G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C22295G 5 g
C222925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C22295G Supplier TCI America Supplier No. C22295G
Only null left

Chemical Identifiers

CAS 79455-63-3
Molecular Formula C7H3Cl2FO
Molecular Weight (g/mol) 192.998
MDL Number MFCD00178746
InChI Key GFNAJZAKJGKJCS-UHFFFAOYSA-N
Synonym 2-chloro-6-fluorobenzoylchloride, 2-chloro-6-fluorobenzene-1-carbonyl chloride, 2-chloro-6-fluoro-benzoyl chloride, 6-chloro-2-fluorobenzoyl chloride, benzoyl chloride, 2-chloro-6-fluoro, acmc-1behg, 2-fluoro-6-chlorobenzoyl chloride, attercop-chm at108371, 2-chloro-6-fluoro benzoyl chloride, 2-chloranyl-6-fluoranyl-benzoyl chloride
PubChem CID 2736547
IUPAC Name 2-chloro-6-fluorobenzoyl chloride
SMILES C1=CC(=C(C(=C1)Cl)C(=O)Cl)F

Specifications

Color Yellow
Quantity 5 g
UN Number 3265
Formula Weight 193.00
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-Chloro-6-fluorobenzoyl Chloride